WHEATON® Diazomethane Generator with ST Clear-Seal™ Grease-Free Joints, DWK Life Sciences
Catalog #: 21518-132
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Description:Diazomethane generator, complete
- Height:190 mm
- Cat. no.:21518-132
- Supplier No.:281135
Specifications
About this item
This Diazomethane Generator is used to generate diazomethane and other gases for gas chromatography, mass spectroscopy and NMR studies using one millimole of Methylnitronitrosoguanidine (MNNG).
- Clear-Seal™ grease-free joints that have never been ground and does not require an 'O' ring
- Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Includes a plastic standard taper joint clamp, a 13-425 screw cap with cut-out top and a PTFE faced rubber liner.
Caution: Please use a safety shield and efficient fume hood when generating diazomethane or similar gases.
Customers Who Bought This Also Bought