About this item
These Cubis® II premium high capacity laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
- Quick and easy, motorized, manual-levelling function, integrated climate sensors, individual sample holders
- Error-free operation with individual QApp workflows and full end-to-end data integrity
- Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
- User Management provides complete access control and configured in accordance with password policy
- Built-in ionizer for electrostatic elimination- electrostatic charges on samples and vessels are removed effectively
- Factory installed basic set of essential weighingapplications
- State of the art TFT touch screen operation with brilliant, readable, display but uncomplex, easy to operate user interface
Climate monitoring ensures that temperature, pressure, and humidity do not negatively affect results. Status center notifications shows all information about balance and environmental conditions like calibration, leveling, temperature, humidity, air pressure and service, centralized in a dashboard. Remove balance stabilization frustrations caused by electrostatic effects with the built-in ionizing technology. It provides modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Flat, stainless steel weight pan with no draft shield for weighing modules.
Specifications
- Catalog No:
- CA76334-416
- CA76334-414
- CA76334-412
- CA76333-942
- CA76334-410
- CA76334-402
- CA76334-406
- CA76334-404
- CA76334-408
- CA76333-816
- CA76333-818
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V