Order Entry
ContactUsLinkComponent
KIMBLE® MICROFLEX® Powder Funnel, DWK Life Sciences
KIMBLE® MICROFLEX® Powder Funnel, DWK Life Sciences
Catalog #: KT747575-0020
Funnels Powder Funnel
KIMBLE® MICROFLEX® Powder Funnel, DWK Life Sciences
Catalog #: KT747575-0020

Specifications

  • Description:
    Powder funnel
  • Overall Height:
    60 mm
  • GPI Thread Finish:
    20–400
  • Overall Width:
    53 mm
  • Supplier No.:
    747575-0020

Specifications

About this item

These powder funnels are useful for the addition of reactants to a variety of flasks.

  • Accessory for Microscale MICROFLEX® threaded kits

Manufactured from 33 expansion, low extractable borosilicate glass.

Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought