KIMBLE® Stoppers for Incubation Flask, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Used for metabolic studies of biological materials. Particularly useful in measuring C¹⁴O² yields following incubation. Also suitable for in vitro incubations, insulin bio-assays, persulfate oxidations, and the distillation of any volatile acid or base.
- The disposable polypropylene center well may be cut off when transferred to a liquid scintillation counting vial
- Rubber stopper provides a double seal and has an off-center hole for the insertion of the center well
- Sidearm flask enables syringe needle entry without disturbing established conditions
Since either an acid or alkali is introduced at the end of the reaction, it is possible to use this easy system with either a phosphate or bicarbonate buffer.
Customers Who Bought This Also Bought