About this item
These MCA series advanced user interface laboratory balances have fully customizable hardware, software, and connectivity including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QApp packages for optional software extension.
- Quick and easy, motorized, auto-levelling function, integrated climate sensors, individual sample holders
- Error-free operation with individual QApp workflows and full end-to-end data integrity
- Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
- User Management provides complete access control and configured in accordance with password policy
- Gesture control allows user to open and close the automatic draft shield, start the ionizer, or send a document
- Maximum capacity between 320 and 14200 g scale interval with a range of 1 to 100 mg
- Easy to use manual glass draft shield
- Removable flat stainless-steel draft shield frame
Climate monitoring ensures that temperature, pressure, and humidity do not negatively impact results. Status center notifications show all information about balance and environmental conditions like calibration, leveling, temperature, humidity, air pressure and service, centralized on a dashboard. Remove balance stabilization frustrations caused by electrostatic effects with the built-in ionizing technology. It provides modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Audit trail and alibi memory with a tamper-protected time-stamped electronic logfile, that allows reconstruction of events related to the creation, modification, and or deletion of records and the system automatically stores weighing data in a ring buffer. The data can be displayed, filtered, and exported in an easy way. It can automatically execute time-controlled actions for backup. The system is able to upload data on a file share or export the data to other systems. Safe data transfer saves all files, together with a calculated MD5 checksum which is stored in the audit trail files. An electronic record should be protected against any manipulation. Built-in ionizer for electrostatic elimination- electrostatic charges on samples and vessels are removed effectively. For all models up to a maximum capacity of 8,2 kg.
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance for all weighing tasks. The Cubis® II range of premium laboratory balances with a maximum load between 2,1 and 70 kg provides the ideal model for every application. It supports automatic time synchronization through Network Time Protocol (NTP). An electronic signature can be used to sign the final report for weighing processes.
Display: Large high end 7" color touch TFT display in 16:9 format with new user interface; Software: Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required); Operation: Activated by touch key, touch-free using IR sensor (draft shield M) or gesture sensor (optional), learning capability.
Draft shield: A- motorized draft shield; E - small glass draft shield; I - motorized draft shield + ionizer; R - stainless steel draft shield frame; U - manual glass draft shield.
Specifications
- Catalog No:
- CA76334-064
- CA76334-062
- CA76334-060
- CA76333-912
- CA76334-058
- CA76334-066
- CA76334-048
- CA76334-056
- CA76334-142
- CA76334-054
- CA76334-052
- CA76334-050
- CA76334-042
- CA76334-040
- CA76334-046
- CA76334-044
- CA76334-030
- CA76334-028
- CA76334-020
- CA76334-018
- CA76334-026
- CA76334-024
- CA76334-022
- CA76334-092
- CA76334-090
- CA76334-088
- CA76334-094
- CA76377-062
- CA76334-086
- CA76334-238
- CA76334-084
- CA76334-236
- CA76334-082
- CA76334-234
- CA76334-080
- CA76333-806
- CA76334-070
- CA76334-068
- CA76334-078
- CA76334-230
- CA76334-076
- CA76334-228
- CA76334-074
- CA76334-072
- CA76333-928
- CA76377-162
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V