Prelabeled Culture Tubes, Disposable, Soda-Lime Glass, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These glass culture tubes feature a permanent vertical ceramic label, fused to the glass, which is easily written on with marker or pencil.
- Permanent marking spots
- Labels are located ³/₄" from the open end and are available in a variety of colors
- All tubes have sturdy, uniform bottoms and consistent lengths
Manufactured from 90 expansion soda-lime glass conforming to USP Type III requirements.
Packaging: 250/pack; 1000/case.
Customers Who Bought This Also Bought