KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
Catalog #: KT953984-0090
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:ULTRA-WARE® conversion base
- Size:90 mm
- Cat. no.:KT953984-0090
- Supplier No.:953984-0090
Specifications
About this item
ULTRA-WARE® conversion base converts the five valve, four valve and filtration caps to use 90 mm filter membranes, providing up to four times faster filtration.
- Requires a 90 mm solvent pickup adapter
The conversion base is manufactured from glass-filled PTFE and is supplied with a coarse porosity (40 to 60 micron porosity) glass support frit and an upper screw clamp.
Customers Who Bought This Also Bought