Precision Balances, Entris® II Essential, Sartorius
Supplier: Sartorius
Certificates
About this item
These Entris® II laboratory balances are ideal for basic weighing tasks.
- Highly accurate results guaranteed via Monolithic weigh cell technology
- Fastest stabilization time in its class using state-of-the-art weighing sensors
- Future-proof USB Type C interface and industry-proof RS232 9-pin interface
- Prevents cross contamination with wipe clean design and easy-to-remove parts
- isoCAL (internal calibration and adjustment) for total assurance of accurate weighing results
- Best repeatability with rectangular weighing pan
- Overload protection with rugged design weighing up to a pre-set amount
- Guaranteed reliability with the self-test '@start'
High chemical resistance ensured using parts made from hard wearing polybutylene terephthalate (PBT), stainless steel and glass. Features 12 built-in applications with GLP/GMP compliant printout or data output. Convenient access to weighing pan and easy-to-remove top and side sliding doors. Draft shield can be removed completely.
Eight built-in languages for international operation - English, German, French, Italian, Spanish, Portuguese, Russian, Polish.
Voltage: 100 to 240 V
Ordering information: Supplied without accessories.
Specifications
- Catalog No:
- CA76334-174
- CA76334-216
- CA76334-172
- CA76334-170
- CA76334-214
- CA76334-168
- CA76334-212
- CA76334-182
- CA76334-180
- CA76333-828
- CA76334-178
- CA76333-830
- CA76334-176
- CA76334-210
- CA76334-208
- CA76334-206
- CA76333-846
- CA76334-196
- CA76334-194
- CA76333-842
- CA76334-192
- CA76333-844
- CA76334-190
- CA76334-204
- CA76334-202
- CA76333-838
- CA76334-166
- CA76334-188
- CA76333-840
- CA76334-164
- CA76334-162
- CA76333-836
- CA76334-186
- CA76334-184
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V