KIMBLE® NMR Tube Washers, DWK Life Scineces
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Design of NMR tube washers uses flexible PTFE tubing to direct a power washstream of wash solvent to clean the inside of NMR tubes.
- Accommodates 7 and 8" tubes
- Vacuum source is required
- All wetted parts are PTFE or borosilicate glass
Aspirator suction provides the driving force to direct the solvent stream into the 5 mm NMR tube. An air stream aids the drying process after the solvent reservoir empties.
Delivery: Complete unit is provided with a one liter reservoir bottle. Supplied with one #8 silicone stopper (for 5 mm NMR tube washer).
Customers Who Bought This Also Bought