MICROFLEX® MidKnight™ XTRA 93-862 Examination Gloves, Ansell Healthcare
Supplier: Ansell Canada
Certificates
About this item
Reliable fentanyl protection that makes it easy to see hazardous materials.
- Tested against both fentanyl and gastric acid (vomit) to simulate extreme, real world overdose situations
- Easy to see when hands have come in contact with potentially hazardous powders because of their dark, black color
- Provides extra protection over the wrist and forearm with an extended cuff
- Made with a soft, flexible nitrile formulation
- Fully textured for a reliable, consistent grip
- Silicone-free
MICROFLEX® MidKnight™ XTRA 93-862 nitrile examination gloves provide comfortable, reliable protection against nasty substances such as opioids. These innovative, extended cuff gloves are tested against both fentanyl and gastric acid (vomit) to simulate extreme, real world overdose situations. The dark black color makes it easy to see hazardous substances while also masking dirt and stains.
Certifications: ASTM D6319, EC 1935/2004, EC 2002/72, EC 2023/2006, EN ISO 374-1:2016, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 374-5:2016, EN 421:2010, FDA21 CFR 177-2600, ISO 9001, ISO 13485, NFPA 1999:2018.
Packaging: Gloves are packaged 100 per dispenser, 10 dispensers per case (1000 gloves per case).
Specifications
- Catalog No:
- 76312-346
- 76312-348
- 76312-350
- 76312-344
- 76312-352
- Finish:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Cuff style:
- Extended
- Extended
- Extended
- Extended
- Extended
- Finger thickness:
- 0.16 mm (6.3 mil.)
- 0.16 mm (6.3 mil.)
- 0.16 mm (6.3 mil.)
- 0.16 mm (6.3 mil.)
- 0.16 mm (6.3 mil.)
- Palm thickness:
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)
- 0.12 mm (4.7 mil)
Specifications
Customers Who Bought This Also Bought