KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Caps and White Rubber Liners, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These sample vials have closed top black phenolic closure with white rubber liners, attached.
- Amber glass vials
Manufactured from 51 expansion borosilicate amber glass conforming to UPS Type I requirements.
Packaging: Packaged in corrugated trays with partitions.
Customers Who Bought This Also Bought