KIMAX® Separatory Funnel, Globe, with PTFE Stopper, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The sharp separations in these funnels are provided for by a narrow bore in the body just above the stopcock.
- Gradually tapering shoulders and a wide neck facilitate cleaning
- Designed from ASTM Specification E1096, Type 2 requirements
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Delivery: Includes standard taper ground glass stopper and a PTFE stopcock.
Customers Who Bought This Also Bought