Vacuum Filtration Adapter with PTFE Stopcock, DWK Life Sciences
Catalog #: KT178130-2942
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- ST Joint:29/42
- Description:Vacuum filtration adapter with PTFE stopcock
- Pluro Stopper:#3
- Fits:150 ml funnel capacity
- Supplier No.:178130-2942
Specifications
About this item
Vacuum filtration adapters with PTFE stopcock is used for reduced pressure filtration using plain stem buchner funnels.
- PTFE stopcock permits control of vacuum source
- Designed with a flange to accommodate a pluro stopper
- Replacement PTFE stopcock plug is 821001 series
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Pluro stopper sold separately.
Customers Who Bought This Also Bought