BioClean DB™ Cleanroom Face Masks, Nitritex
Supplier: Ansell Healthcare
Certificates
About this item
The BioClean DB™ Pouch-style Facemaskd feature a large breathing chamber for increased wearer comfort. Made from cleanroom compatible materials facemasks are available with or without an integrated neck guard to provide additional coverage, reducing the risk of cross-contamination.
- Non-woven spunbonded polyester (hygroscopic) inner facing layer
- Meltblown polypropylene filter layer
- Non-woven spunbonded polyester (hydrophobic) outer facing layer
- Tubular knitted polyurethane Spandex yarn headloops fastenings
- Non-woven spunbonded polyester neck guard
- Plastic coated steel nose band
High bacterial and particle filtration efficiency with extra a long neck guard. It consists of large breathing chamber, ultrasonically sealed edges, and fully enclosed malleable nose-band.
Certifications: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1992.
Packaging: 50 pieces per sealed inner PE bag, six inner bags per sealed outer PE bag, one outer bag per lined carton (300 pieces).
Specifications
- Catalog No:
- CA75972-528
- Material:
- PE