Wizard Series PVC/Nylon/PVC Rain Jackets with Detachable Hood, MCR Safety
Supplier: MCR Safety
Certificates
About this item
This wizard series coat features a 0.28 mm double coating of PVC, with tough nylon scrim material layered in between.
- Equipped with detachable drawstring hood
- Take up snaps on the wrists
- Storm flap under snap front closure
- Rip stop scrim material provides excellent resistance to snags and tears
Nylon scrim has excellent rip and puncture resistance. This yellow two-piece jacket is a waist length option with a detachable drawstring hood and storm flap under a snap up front closure. Limited flammability, this garment is not intended for use as protection against ARC flash or flash fire.
Specifications
- Catalog No:
- 76341-064
- 76340-874
- 76341-150
- 76341-008
- 76341-114
- 76340-952
- 76341-038
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC