Wizard Series PVC/Nylon/PVC Knee Length Coats with Detachable Hood, MCR Safety
Supplier: MCR Safety
Certificates
About this item
This wizard series coat features a 0.28 mm double coating of PVC, with tough nylon scrim material layered in between.
- Knee length design
- Detachable drawstring hood
- Rip stop scrim material provides excellent resistance to snags and tears
The nylon scrim provides excellent rip and puncture resistance. This yellow two-piece coat has a 49" total length (knee length) and includes a detachable hood and storm flap under a snap up front closure. Limited flammability, this garment is not intended for use as protection against ARC flash or flash fire.
Specifications
- Catalog No:
- 76341-134
- 76341-132
- 76341-526
- 76341-328
- 76341-604
- 76341-210
- 76341-452
- 76341-324
- 76341-198
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC