Big Jake® PVC / Nomex® Flame Resistant Rain Jackets, MCR Safety
Supplier: MCR Safety
Certificates
About this item
The Big Jake® is a flame resistant jacket made of PVC/Nomex® material that will keep wearers dry and safe from certain FR hazards.
- Sturdy zipper front with large storm flap
- Cape ventilated back with D-Ring access
- 2" silver FR reflective stripes
- Two front patch pockets
- Adjustable/attached drawstring hood
- Adjustable hook and loop wrist
Jackets feature a sturdy zipper front, oversized attached adjustable drawstring hood, adjustable hook and loop wrist closures, two front patch pockets, and a cape vented back with D-Ring access. The fluorescent lime background material and ensure high visibility protection. These garments also meet the ASTM F1891 standard specification for arc and flame resistant rainwear.
Certifications: Meets CSA Z96 Class 2 Level 2 requirements. When worn in conjunction with BJ38BP bib pants, entire suit meets CSA Z96-15 Class 3 standards; Meets ANSI 107 Class 3 Type R high visibility standards; These garments also meet the ASTM F1891 standard specification for arc and flame resistant rainwear; HRC2 / CAT2, arc thermal performance value (ATPV) 8.9 cal/cm².
Specifications
- Catalog No:
- 76341-910
- Garment application:
- Industrial, Fire protection
- Risk area:
- HiVis
- Gender:
- Unisex
- Closure type:
- Zip closure
- Fabric:
- PVC