Aspirator
Catalog #: 470320-974
Supplier: United Scientific Supplies
Specifications
- Description:Aspirator
- Cat. no.:470320-974
Specifications
About this item
Strong plastic aspirator for vacuum filtration and other lab applications.
- Prevents flooding and back flow
Designed to prevent any flooding, back-flow, or turbulence. Includes 5.9" (15 cm) hose. The top nozzle has an outer diameter of 1.5 cm. The side nozzle can accommodate hoses from 0.5 to 1.0 cm inner diameter.
Customers Who Bought This Also Bought