To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Linearity FD general chemistry for Roche Systems
- Size:10×5 ml
- Cat. no.:76373-814
- Stability:3 days when stored at 2...8 °C
- Analyte:Albumin, Alkaline Phosphatase, ALT, Amylase, AST, BUN
Calcium, Chloride, CO2, Creatine Kinase (CK), Creatinine, Gamma-GT, Glucose, Iron, Lactate (Lactic Acid), LDH, Lipase, Magnesium, Phosphorus, Potassium, Protein- Total, Sodium, Uric Acid - Type:Freeze dried
Specifications
About this item
Universal linearity general chemistry, linearity general chemistry for Beckman AU, and linearity FD general chemistry for Roche Systems are assayed quality control materials consisting of five levels.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.