Valves, High Vacuum, Protected, 90° T metering, PTFE , Chemscience
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Viton® O-rings on PTFE shaft are protected with precision PTFE wipers.
- The plug has a machined-in integral PTFE tip O-ring to make the primary seal
- This design provides an excellent vacuum tight seal
- Does not expose any material other than PTFE to chemicals
The glass portion has two side arms sealed to the barrel at a 90° angle to one another.
Customers Who Bought This Also Bought