Valves, High Vacuum, O-Ring, 90°, PTFE, Chemscience
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
The non-rotating PTFE plug has three O-rings.
- With two 9 mm O.D. side arms
- Sealed to the barrel at a 90° angle to one another
The tip O-ring making the primary seal on the precision formed tapered seat.
Customers Who Bought This Also Bought