Gas Dispersion Tubes, Fritted, ChemScience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
These gas dispersion tubes are available straight or with a 15° or 90° angle.
- Stem has O.D. of 8 mm and length of 150 mm
The 15° angle permits tube to be inserted into vessels where a large opening is not available. The 90° angle permits bubbles to be spread over a large area.
Customers Who Bought This Also Bought