Order Entry
ContactUsLinkComponent
KIMBLE® Separatory Funnels, Squibb Type, Pear Shaped, DWK Life Sciences
KIMBLE® Separatory Funnels, Squibb Type, Pear Shaped, DWK Life Sciences
Catalog #: KT636280-2000
Funnels Separatory [standard taper] 24/40 Joints with PTFE Plug and Stopper
KIMBLE® Separatory Funnels, Squibb Type, Pear Shaped, DWK Life Sciences
Catalog #: KT636280-2000

Specifications

  • Capacity:
    2000 ml
  • Description:
    Seporatory funnel
  • Height:
    437 mm
  • Cat. no.:
    KT636280-2000
  • ST Joint Size:
    24/25
  • Supplier No.:
    636280-2000

Specifications

About this item

KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint

  • Standard taper 24/40 bottom joints
  • Manufactured from 33 expansion, low extractable borosilicate glass

Squibb-type with PTFE plug-style stopcocks. Replacement stoppers are KT850400-2425.

Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought