KIMBLE® Spargers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These U-shaped spargers are designed for use on the Teledyne Tekmar® 2000 and 3000 purge and trap concentrators when doing EPA method 603 and using either an AQUATek 50 or ALS 2050 vial autosampler.
- Manufactured from 33 expansion, low extractable borosilicate glass
The introduction stem is located on the foam trap, allowing a sample heater to be placed around the glassware as required by the EPA method.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought