KIMBLE® Snyder Distillation Column, DWK Life Sciences
Catalog #: KT503100-0003
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Distillation column
- Cat. no.:KT503100-0003
- ST Joint Size:24/40
- Supplier No.:503100-0003
Specifications
About this item
Improved design speeds evaporation and reduces hold-up within the column.
- Recoveries are excellent, example with pesticides such as lindane and parathion in petroleum ether or ethyl ether
- Higher boiling solvents also may be used to good advantage
- Each chamber is approximately 50 mm long
- Manufactured from 33 expansion, low extractable borosilicate glass
Kimble® improved snyder distillation column with 3-chamber and 24/40 [ST] joints.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought