KIMBLE® Cyanide Distillation Apparatus With Separate Scrubber and Absorber, DWK Life Sciences
Catalog #: KT479120-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:1000 ml
- Description:Distillation apparatus
- Cat. no.:KT479120-0000
- ST Joint Size:19/38
- Supplier No.:479120-0000
Specifications
About this item
Cyanide distillation 19/38 apparatus with separate scrubber and absorber.
- Samples react with acid to convert cyanide to HCN
- The first bubbler acts as a scrubber to remove sulfides
- Cyanide is absorbed in the second bubbler
- Design features compact standard taper 19/38 joints
- Manufactured from 33 expansion, low extractable borosilicate glass
Apparatus for removing interfering substances in quantitative analysis of industrial waste waters, saline and reagent grade water.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought