To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Laminin subunit alpha-1; Laminin subunit beta-1; Laminin subunit gamma-1
- Clonality:Polyclonal
- Gene ID:LAMA1; LAMB1; LAMC1
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:LAMA1; LAMB1; LAMC1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Form:Lyophilized
- Storage temperature:Store lyophilised antibody at 2…8° C. After reconstitution divide into aliquots and store at -20° C for long-term storage. Store at 2…8° C short-term (up to 4 weeks). Avoid repetitive freeze/thaw cycles.
- Shipping temperature:Room (ambient) temperature
- Immunogen:Laminin-111 isolated from mouse EHS cells
- Purification:Affinity purified
- Cat. no.:76334-042
Specifications
About this item
Binding to cells via a high affinity receptor, laminin is thought to mediate the attachment, migration and organisation of cells into tissues during embryonic development by interacting with other extracellular matrix components. Ref: uniprot.org
This antibody recognizes laminin isotypes alpha-1 (440 kDa), beta-1 (220 kDa) and gamma-1 (220 kDa). It also binds laminin binding protein at 120 kDa which always co-expresses with laminin.
Applications: Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: LAMA1; LAMB1; LAMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, rat