Diamond® RealSeal™ Bottles, Narrow Mouth, Polypropylene, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
- Suitable for autoclaving at 121 °C (including cap)
- Improved chemical resistance and clarity
- Resistant to most corrosives and suitable for a wide variety of laboratory uses
- Approved for use from –40 to +121 °C
- Medical grade resins meet demanding requirements
- Laboratory bottle quality and performance with savings associated with economy bottles
- Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These narrow mouth bottles improve pouring of liquid contents making them perfect for storing and shipping liquids.
Customers Who Bought This Also Bought