Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Lunar caustic,Silver mononitrate
Formula:AgNO₃
MW:169.87 g/mol
MDL Number:MFCD00003414
CAS Number:7761-88-8
:Storage Temperature: Ambient
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ100
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 210 °C
:Flash Pt:
:Flash Pt:
:M.W. 169.87 g/mol
EINECS:231-853-9
:210
:444
UN:1760
:8
:III
:GHS05
:GHS09
Storage Temperature:Ambient
:MQ100
:12,08661
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Supelco®, Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution:
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_Silver nitrate solution-0.1M.pdf Stability data sheet Titripac_109081.pdf
Product specification
Customers Who Bought This Also Bought