Titriplex® IV solution c (DCTA Na₂) = 0.1 mol/l, Titripur® for metal titration, Supelco®
Catalog #: CA1.08447.1000
Supplier: MilliporeSigma
CAS Number: 5786-78-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Size:1 L
- Cat. no.:CA1.08447.1000
- Supplier No.:1.08447.1000
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
- The concentration is determined by volumetric titration and refers to 20°C.:
- The certified value is traceable to a primary standard from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 682 zinc) by means of volumetric standard zinc (Art. 1.02409).:
- This volumetric standard is certified by the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO 17025.:
- The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%. All measurements are carried out in the laboratory of Merck KGaA, Darmstadt, Germany.:
Specification Test Results
About this item
Synonyms: CDTA disodium salt
Density:1.02 g/cm3 (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ100
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 390.3 g/mol
:MQ100
Formula:C₁₄H₂₀N₂Na₂O₈
MW:390.3 g/mol
CAS Number:5786-78-7
Customers Who Bought This Also Bought