BioClean-D™ Sterile Coveralls with Hood and Integrated Boots, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
The BioClean-D™ coverall with hood and integrated boots is a disposable garment offering comfort and head-to-toe protection.
- Anti-static, lightweight, low-linting CleanTough™ material construction
- Thumb loops to ensure a secure hold
- Front zip with sealable cover and protective flap
- Elasticated back, cuffs, and ankles
- Slip resistant soles
- Excellent ESD properties
- Latex-free
The integrated boots feature slip-resistant polyurethane soles to ensure every step can be taken with confidence. ISO Class 4 cleanroom compatible. These sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739, EN1149-5, CATEGORY III, Sterilized in accordance with ISO 11137-1:2006.
Specifications
- Catalog No:
- CA75982-206
- CA75982-214
- CA75982-212
- CA75982-204
- CA75982-202
- CA75982-210
- CA75982-208
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
Specifications
Related Information
Certificates
Search by Lot Number75982-202_22-1849 75982-202_22-2437 75982-204_19-0809 75982-208_22-2330 75982-210_22-2331
Conformity declaration
More
Customers Who Bought This Also Bought