BioClean-D™ Sterile Coveralls with Hood, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
The BioClean-D™ coverall with hood is a disposable garment.
- Lightweight CleanTough™ material
- Thumb loops to ensure a secure hold
- Front zip with sealable cover and protective flap
- Elasticated hood, back, cuffs, and ankles
- Excellent ESD properties
- Latex-free
The anti-static, low-linting CleanTough™ material provides comfort and protection from a range of chemicals. Lightweight yet durable garments feature bound seams with single needle stitching. ISO Class 4 cleanroom compatible. These sterile garments are gamma irradiated to an Sterility Assurance Level (SAL) of 10⁻⁶.
Certifications: ASTM F739, EN ISO 13688:2013, EN 1 ASTM F739, EN ISO 13688:2013, EN 14325.
Specifications
- Catalog No:
- CA75982-272
- CA75982-270
- CA75982-276
- CA75982-274
- CA75982-278
- CA75982-282
- CA75982-280
- CA75982-284
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound