BioClean-D™ Non sterile Coveralls with Hood, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles, and thumb loops to ensure a secure hold.
- Lightweight CleanTough™ material
- Thumb loops to ensure a secure hold
- Front zip with sealable cover
- Elasticated hood, back, cuffs, and ankles
- Excellent ESD properties
- Latex-free
The anti-static, low-linting CleanTough™ material provides comfort and protection from a range of chemicals. Lightweight yet durable garments feature bound seams with single needle stitching. ISO Class 4 cleanroom compatible.
Certifications: ASTM F739, EN ISO 13688:2013, EN 1 ASTM F739, EN ISO 13688:2013, EN 14325.
Specifications
- Catalog No:
- CA75972-550
- CA75972-656
- CA75972-554
- CA75972-552
- CA75972-556
- CA75972-560
- CA75972-558
- CA75972-562
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound