Order Entry
ContactUsLinkComponent
Free Cholesterol E, Wako
Free Cholesterol E, Wako
Catalog #: CA11001-650
Supplier:  Wako Life Sciences
CAS Number:  
Free Cholesterol E, Wako
Catalog #: CA11001-650
Supplier:  Wako Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Free Cholesterol E
  • Size:
    2 x 75 mL, 1 x 10 mL (kit)
  • Storage temperature:
    2-10°C
  • Cat. no.:
    CA11001-650
  • Supplier No.:
    435-35801

Specifications

About this item

The Wako Free Cholesterol E is an in vitro assay for the quantitative determination of free cholesterol in serum.

Method: Enzymatic

Composition
Buffer Solution: 2 x 75 mL
Color Reagent: 2 x for 75 mL
Standard Solution: 1 x 10 mL

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought