LD-100 System Frocks, 469 Series, WORKLON®
Supplier: Superior Uniform
Certificates
About this item
Reusable frocks feature a snap closure and set-in sleeves.
- Adjustable snaps at wrist and collar
- Color-coded hang loop denotes size
- Unisex design
The 100% polyester taffeta construction allows for use in a variety of industries requiring particulate control and resistance to chemicals and abrasion. These frocks are engineered to meet up to Class 1000 (ISO 6) cleanroom requirements.
Specifications
- Catalog No:
- 10827-196
- 10827-200
- 10827-198
- 10827-204
- 10827-202
- 10830-608
- 10830-610
- 10827-194
- 10827-192
- Garment application:
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Food, Cleanroom, Chemical protection, General Purpose, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester