Cart Covers, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Cart covers help prevent airborne contaminants from resting on items stored on carts. Cart covers are also a good means of deterring pilferage.
- Lightweight and easy to handle
- Reinforced corners for added life
- Uncoated covers: 100% polyester, machine-washable
- Transparent vinyl allows for ease of viewing cart contents
- Many sizes available to fit virtually any application
Coated covers made of coated fabric vinyl nylon, which is waterproof, fire-retardant, and resists tearing, odor, mildew, and flames. ESD, these cart covers are electrically conductive, antibacterial, antistatic, flame retardant, waterproof, tear resistant, reinforced vinyl laminate fabric. Includes a flap on one side, brass zipper and paperwork pouch.
Customers Who Bought This Also Bought