Flasks, Heavy Wall, Round Bottom, Single Neck With Hooks, ChemScience
Supplier: CHEM SCIENCE INC CA
Certificates
Certificates
About this item
Heavy wall design.
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Ethanol denatured, HiPerSolv CHROMANORM® for HPLC (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
$536.97 to $1,146.83
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$146.85 to $576.51
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$814.96