KleenGuard™ A30 Breathable Splash and Particle Protection Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
Certificates
About this item
The KleenGuard™ A30 breathable splash and particle protection coveralls feature breathable fabric and reflex design, which offers excellent fit (with elastic back) and freedom of movement.
- Designed to protect against dirt, grime, and light splashes
- Made with breathable barrier fabric
- Fabric features an intricate web of micro-fibers that filter out many water-based liquids and dry particulates
- Reflex coverall design offers excellent fit and freedom of movement (also includes elastic back)
- Skid-resistant boots provide up to 68% improvement in traction on wet and dry surfaces when compared to former boot material
The material is strong and abrasion resistant, and filters out many water-based liquids and dry particles. Other A30 features include a seamless front (for primary exposure area), 1" zipper flap, hood, elastic wrists.
These coveralls protect against hazardous dry particles, aerosols, and non-hazardous light liquid splashes, and are perfect for environments such as pharmaceutical research, aerospace, maintenance, and general manufacturing.
Certifications: Passes NFPA 99 for antistatic materials.
Specifications
- Catalog No:
- 76300-934
- Model:
- A30
- Fabric:
- SMS
- Anti-static:
- Yes
- Garment application:
- General Purpose, Laboratory
Specifications
Customers Who Bought This Also Bought