Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis), VWR® ACS, Reagent Grade for histology
Catalog #: 89370-088
Supplier: VWR International
Product available on GSA Advantage®
CAS Number: 1330-20-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 gal.
Specifications
Specification Test Results
- Chemical Assay (GC), (Xylene Isomers & Ethyl Benzene):98.5% min.
- Paraxylene ( p-xylene):Reported per lot
- Metaxylene (m-xylene):Reported per lot
- Orthoxylene (o-xylene):Reported per lot
- Ethyl Benzene:25% max.
- Boiling Range:137-143 °C
- Color (APHA) :10 max.
- Appearance:Clear
- Residue After Evaporation:10 ppm max.
- Substances Darkened by H₂SO₄:To Pass
- Sulphur Compounds:1 ppm max.
- Water:0.05% max.
- Specific Gravity @ 15.56°C :0.865 - 0.875
- Benzene Content:0.01% max.
- Toluene Content:0.5% max.
- Total C9 Aromatics:Reported per lot
- Non-Aromatics:0.30% max.
- Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis), VWR® ACS, Reagent Grade for histology:
Specification Test Results
About this item
Synonyms: Dimethylbenzene
:Storage Temperature: Ambient
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5
Flash Pt:25 °C
:Boiling Pt:
:Boiling Pt: 140 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –34 °C
:Flash Pt:
:Flash Pt: 25 °C
:M.W. 106.17 g/mol
Merck Index:12,10214
Formula:C₆H₄(CH₃)₂
MW:106.17 g/mol
CAS Number:1330-20-7
EINECS:215-535-7
:140 °C (1013 hPa)
UN:1307
:3
:III
:25 °C
:GHS02
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00077264
Customers Who Bought This Also Bought