2-Propanol ≥99.0%, VWR® ACS, Reagent Grade for histology
Catalog #: 89370-086
Supplier: VWR International
Product available on GSA Advantage®
CAS Number: 67-63-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 gal.
Specifications
Specification Test Results
- USP Assay (by GC) (corrected for water):min. 99.0%
- ACS Assay (by GC) (corrected for water):min. 99.5%
- USP Identification A:To Pass
- USP Identification B:To Pass
- ACS Color (APHA):max.10
- ACS Appearance :Clear Liquid
- ACS Solubility in Water:Solution as clear as equal volume of water
- ACS Residue After Evaporation:max.
- USP Limit of Non-Volatile Residue:max. 2.5 mg/50 ml
- USP Limit of Volatile Impurities :
- Diethyl Ether:max. 0.1%
- Acetone:max. 0.1%
- Diisopropyl Ether:max. 0.1%
- n-Propyl Alcohol:max. 0.1%
- 2-Butanol:max. 0.1%
- Total :max. 1.0%
- ACS Titrable Acid or Base:max. 0.0001 meq/g
- USP Acidity:NMT 0.70 mL of 0.02N NaOH required for neutralization
- ACS Water:max. 0.2%
- USP Water Determination :max. 0.5%
- ACS Carbonyl Compounds 0.002% max. each of propionaldehyde or acetone :max. 0.002%
- USP Refractive Index @ 20°C:1.376-1.378
- USP Specific Gravity @25°C:0.783-0.787
- 2-Propanol ≥99.0%, VWR® ACS, Reagent Grade for histology:
Specification Test Results
About this item
Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol
:2-Propanol
:Storage Temperature: Ambient
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
Flash Pt:12 °C
:Boiling Pt:
:Boiling Pt: 82 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 60.1 g/mol
MW:60.1 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00011674
CAS Number:67-63-0
EINECS:200-661-7
:82 °C (1013 hPa)
UN:1219
REACH:01-2119457558-25
:3
:II
:12 °C
:GHS02
:GHS07
Formula:(CH₃)₂CHOH
Customers Who Bought This Also Bought