About this item
The KleenGuard™ A45 liquid and particle protection coveralls for surface prep and paint are designed to protect workers where hazardous or non-hazardous contaminants may be present.
- Multiple styles are available
- Made of microporous film laminate with a water-resistant zipper front
- REFLEX coverall design offers excellent fit and freedom of movement, so workers won't need to modify them to do their job
- Coveralls with a hood are designed to fit a respirator
- Protects against chromate dust (10%) and isocyanates (40% concentration), hazardous materials often used in industrial paint jobs
- Skid-resistant boots provide up to 68% improvement in traction on wet and dry surfaces when compared to former boot materials
The KleenGuard™ A45 liquid and particle protection coveralls for surface prep and paint are designed to move with workers where hazardous or non-hazardous contaminants may be present. Each protective suit is made with lightweight fabric, allowing for improved comfort without sacrificing protection. They are not made with natural rubber latex and are silicone-free, and they feature serged seams and a water resistant zipper.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76300-892
- 76300-936
- 76300-894
- 76300-938
- 76300-896
- 76300-898
- 76300-900
- 76300-902
- CA10789-204
- CA10789-224
- CA10789-222
- CA10789-212
- CA10789-210
- CA10789-208
- CA10789-206
- CA10789-220
- CA10789-218
- CA10789-216
- CA10789-214
- CA10789-226
- Model:
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- A45
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial