Acetonitrile ≥99.9% (by GC) for DNA synthesis, Burdick & Jackson™
Catalog #: CABJ34442-2.5L
Supplier: Burdick & Jackson
New Product
CAS Number: 75-05-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:2.5 l
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Water (Karl Fischer):Max. 0.001 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
- Burdick & Jackson™, Acetonitrile ≥99.9% (by GC) for DNA synthesis:
Specification Test Results
About this item
Synonyms: Methyl cyanide,Ethyl nitrile,Cyanomethane,Ethanenitrile
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:-46 °C
Density:0.7857 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:6 °C
:Boiling Pt:
:Boiling Pt: 82 °C (1.013 hPa)
:Melting Pt:
:Melting Pt: -46 °C
:Flash Pt:
:Flash Pt: 6 °C
:M.W. 41.05 g/mol
:Acetonitrile
Formula:H₃CCN
MW:41.05 g/mol
EINECS:200-835-2
:82 °C (1.013 hPa)
Storage Temperature:Ambient
MDL Number:MFCD00001878
CAS Number:75-05-8
UN:1648
REACH:01-2119471307-38
:3
:II
:6 °C
:GHS02
:GHS07
Customers Who Bought This Also Bought