ESD Inspection Gloves, Desco
Supplier: Desco
Certificates
About this item
ESD Inspection Gloves are made to protect the operator's hands from cuts caused by circuit board edges, leads, and other sharp objects.
- Protective carrier yarn
- Seamless fine gauge knit
- Non-scratch touchscreen compatible
- Durable construction
Dissipative (1×10E5 to <1×10E8 ohms) Thunderon® Fibers reduce charge generation and bond to a grounded operator. Protective carrier yarn protects the operator's hands from cuts caused by circuit board edges, leads, and other sharp objects. Absorbs oils and other contaminants produced by operator's skin.
Seamless fine gauge knit is designed for all-day wear, allowing easy hand movement and a high level of tactile sensitivity. Wearer can operate touch screen monitors and other devices. Durable construction will retain ESD and other properties for 25 washings or approximately one year.
Specifications
- Catalog No:
- 10199-484
- 10199-488
- 10199-486
- 10199-490
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Anti-static:
- Yes
- Yes
- Yes
- Yes