Humidity Indicator Card, Desco
Catalog #: 89131-360
Supplier: Desco
Product available on GSA Advantage®
Specifications
- Description:Humidity Indicator Card
- Cat. no.:89131-360
- Supplier No.:13870
Specifications
About this item
Reversible Humidity Indicator Cards for vapor protection measure the level of humidity that the contents in package are exposed.
- For use with Statshield® ESD Shielding Moisture Barrier Bags
- Humidity indicator spots are reversible
- 100 cards per can
- Made in the United States of America
Cards measure the level of humidity that the contents of the packaging have been exposed to. They can be used with Statshield® ESD Shielding Moisture Barrier Bags to provide protection for moisture sensitive items such as surface mount devices. Reversible design allows the card to be reused, providing better value.
Certifications: Meets MIL-I-8835A And JEDEC J-STD-033D (Type 1) Table 3-1 Dry Packing requirements. Lead-Free, RoHS compliant.
Customers Who Bought This Also Bought