Statfree® High Density Conductive Foam, Desco
Catalog #: 89131-086
Supplier: Desco
Product available on GSA Advantage®
Specifications
- Description:Statfree® high density conductive foam
- Cat. no.:89131-086
- Dimensions:0.75×24×36"
- Supplier No.:12550
Specifications
About this item
High Density Conductive Foam is antistatic and low tribocharging, which minimizes charge generation.
- Antistatic, low tribo charging
- Dissipative and corrosion resistant
- Uniform protection
- Volume Resistance: 10E3 – 10E5 ohms
- Surface Resistance: 10E3 – 10E5 ohms
- Made in the United States of America
Antistatic, low tribocharging minimizes charge generation; material will conduct charges if grounded. Corrosion resistant design is ideal for long-term storage. Uniform protection will not affect the solderability of device leads.
Certifications: Lead-free, RoHS compliant.
Customers Who Bought This Also Bought