Dissipative Label Dispenser, Desco
Catalog #: 10199-584
Supplier: Desco
Product available on GSA Advantage®
Specifications
- Description:Dissipative label dispenser
- Cat. no.:10199-584
- Supplier No.:06767
Specifications
About this item
Dissipative Label Dispenser helps peel away backing from label. Designed for use with labels up to 2" wide.
- Volume Resistance: Rv 1×10E6 to < 1×10E9 ohms per ANSI/ESD STM11.12
- Made in the United States of America
Designed for used in an ESD protected area. Place on a grounded ESD work surface to ground. Label rolls must have a core to work with the label dispenser.
Ordering information: Item ships unassembled.
Customers Who Bought This Also Bought