Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
Catalog #: 470232-940
Supplier: WALLCUR, LLC.
Specifications
- Capacity:5 ml
- Description:Traning Fentanyl Ampules, 5 ml
- Cat. no.:470232-940
Specifications
About this item
Medication admistration simulation and training.
- For training purposes only
Practi-Fentanyl™ 5 ml Ampule for clinical training, accurately simulates 250 mcg/5 ml (50 mcg/ml) (0.05 mg/ml) of the synthetic opioid analgesic, Fentanyl. Each of it comes as a labeled 5 ml ampule containing distilled water.
Caution: Not for human or animal use.
Customers Who Bought This Also Bought