Audible Continuity Tester, Desco Industries
Catalog #: 15431-820
Supplier: Desco
Specifications
- Description:Audible Continuity Tester
- Cat. no.:15431-820
- Supplier No.:35135
Specifications
About this item
Audible continuity testers are used to confirm electrical circuit continuity, even in bright environments.
- Range: 0 to 50 ohms
- Probe: 3 volts at 15 ma maximum
- Strong audible 2 khz tone clearly confirms electrical circuit continuity, even in bright environments
Delivery: Includes 2 AAA batteries.
Customers Who Bought This Also Bought