Stainless Steel Probe, Desco Industries
Supplier: Desco
Certificates
About this item
The 5.5" stainless steel probe is ideal for working in ESD protective areas.
- Ergonomic hex handle
- Non-magnetic, conductive 300 series stainless steel
- Stainless steel won’t corrode - no problem in working with magnetized parts
- Any charge can be grounded when placed on ESD protected work surface
Made in the United States of America superior quality.
Delivery: The stainless steel probe kit includes one each of the straight, hooked, angled, and curved probe versions.
Customers Who Bought This Also Bought