Specifications
- Description:Dual Luer Swivel Fittings
- Cat. no.:89134-252
- Gauge:22G
- Supplier No.:QCS-D
Specifications
About this item
The Quick Connect™ Swivel provides Luer-Lock fittings for trouble-free connection to extension lines and tethers, making connecting simple, fast, and easy.
- Fits all standard luer fittings
- Double Luer and Single Luer models available
- EtO exposed
Add injection caps, valves, or stopcocks to simplify dosing or sampling procedures. Part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether and Quick Connect™ Harness for tethered small animal infusion studies.